Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763411

Sigma-Aldrich

Dichloro(norbornadiene)platinum(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8Cl2Pt
CAS Number:
Molecular Weight:
358.12
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: platinum
reagent type: catalyst

mp

186-218 °C (decomposition)

Storage temp.

2-8°C

SMILES string

Cl[Pt]Cl.C1[C@H]2C=C[C@@H]1C=C2

InChI

1S/C7H8.2ClH.Pt/c1-2-7-4-3-6(1)5-7;;;/h1-4,6-7H,5H2;2*1H;/q;;;+2/p-2/t6-,7+;;;

Inchi Key

AYGLNRVTDUMGOQ-HBJFUXGYSA-L

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service