Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76340

Sigma-Aldrich

4-Amino-1-butanethiol 4-methoxytrityl thioether polymer-bound

extent of labeling: 0.3-1 mmol/g loading

Synonym(s):

4-(4-Methoxytritylthio)butylamine polymer-bound, 4-Aminobutyl 4-methoxytrityl thioether polymer-bound

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352202
NACRES:
NA.22

form

lumps

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

extent of labeling

0.3-1 mmol/g loading

matrix

(crosslinked with 1% DVB)

particle size

200-400 mesh

application(s)

peptide synthesis

storage temp.

2-8°C

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service