Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763276

Sigma-Aldrich

Bis(N,N-dimethyl-β-alaninato)palladium(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O4Pd
CAS Number:
Molecular Weight:
338.70
UNSPSC Code:
12352300
PubChem Substance ID:

assay

97%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

152-172 °C

storage temp.

2-8°C

SMILES string

CN(C)CCC(=O)O[Pd]OC(=O)CCN(C)C

InChI

1S/2C5H11NO2.Pd/c2*1-6(2)4-3-5(7)8;/h2*3-4H2,1-2H3,(H,7,8);/q;;+2/p-2

InChI key

ZXNOSELUGCGIPS-UHFFFAOYSA-L

Application

Phosphine free palladium catalyst for Suzuki Cross-Couplings

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xin Cui et al.
The Journal of organic chemistry, 72(24), 9342-9345 (2007-11-02)
N,O-Bidentate compounds were systematically evaluated as phosphine-free ligands for Pd-catalyzed C-C bond-formation reactions through kinetic measurements. Pd(quinoline-8-carboxylate)2 was identified as one of the most efficient, yet still low-priced, phosphine-free catalysts for Heck as well as Suzuki reactions of unactivated aryl
Cui, X.; et al.
Synthesis, 3, 393-393 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service