Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

763179

Sigma-Aldrich

{2-[(α-Methyl)diphenylphosphinomethyl]ferrocenyl}palladium(II) acetate

Synonym(s):

{2-[1-(Diphenylphosphino)ethyl]ferrocenyl}palladium(II) acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H25FeO2PPd
CAS Number:
Molecular Weight:
562.71
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

145-185 °C (decomposition)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.CC([C]2[CH][CH][CH][C]2[Pd]OC(C)=O)P(c3ccccc3)c4ccccc4

InChI

1S/C19H17P.C5H5.C2H4O2.Fe.Pd/c1-16(17-10-8-9-11-17)20(18-12-4-2-5-13-18)19-14-6-3-7-15-19;1-2-4-5-3-1;1-2(3)4;;/h2-10,12-16H,1H3;1-5H;1H3,(H,3,4);;/q;;;;+1/p-1

InChI key

GIYKFWJEBAVHNF-UHFFFAOYSA-M

pictograms

Health hazardExclamation mark

signalword

Warning

Hazard Classifications

Carc. 2 - Repr. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ping He et al.
Organic letters, 9(2), 343-346 (2007-01-16)
Anionic four-electron donor-based palladacycle-catalyzed 1,4-additions of arylboronic acids with alpha,beta-unsaturated ketones and 1,2-additions of arylboronic acids with aldehydes and alpha-ketoesters are described. Our study demonstrated that palladacycles were highly efficient, practical catalysts for these addition reactions. The work described here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service