Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762911

Sigma-Aldrich

S-(4-Azidobutyl)thioacetate

97%

Synonym(s):

S-(4-Azidobutyl) ethanethioate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11N3OS
CAS Number:
Molecular Weight:
173.24
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

liquid

refractive index

n20/D 1.501

density

1.100 g/mL at 25 °C

SMILES string

CC(=O)SCCCCN=[N+]=[N-]

InChI

1S/C6H11N3OS/c1-6(10)11-5-3-2-4-8-9-7/h2-5H2,1H3

InChI key

IFUADVSNNZKOGP-UHFFFAOYSA-N

General description

S-(4-Azidobutyl)thioacetate is used as a precursor in the formation of organic self-assembled monolayers (SAMs) on metals. They are chemisorbed onto metal surfaces to form self-assembled monolayers with a high degree of structural order. They are often used as a molecular template to form functional nanostructures.[1]

Application

Used in the formation of Self-Assembled Monolayers on gold electrodes to modify its work function. [1][2]

Features and Benefits

High chemical stability in air or solution. Prevents undesirable oxidation of thiol groups in oxygen containing atmosphere. Forms homogeneous interface structure in SAMs.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

>230.0 °F

flash_point_c

> 110 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Thioacetate-Functionalized Self-Assembled Monolayers on Au: Toward High-Performance Ion-Selective Electrode for Ag
Jin, J., Zhou, W. J., Chen, Y., Liu, Y. L., Sun, X. Q., & Xi, H. T.
Bull. Korean Chem. Soc., 35(2), 601-601 (2014)
Formation and Structure of Self-Assembled Monolayers of Octylthioacetates on Au (111) in Catalytic Tetrabutylammonium Cyanide Solution
Park, T., Kang, H., Choi, I., Chung, H., Ito, E., Hara, M., & Noh, J.
Bull. Korean Chem. Soc., 30(2), 441-441 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service