Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762687

Sigma-Aldrich

Pentathiodiphosphorus(V) acid-P,P′-bis(pyridinium betaine)

Synonym(s):

1,1′-[Thiobis(mercaptophosphinothioylidene)]bispyridinium, bis(inner salt), P4S10-Pyridine complex

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10N2P2S5
CAS Number:
Molecular Weight:
380.47
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

mp

190-210 °C

SMILES string

[S-]P(=S)(SP([S-])(=S)[n+]1ccccc1)[n+]2ccccc2

InChI

1S/C10H10N2P2S5/c15-13(16,11-7-3-1-4-8-11)19-14(17,18)12-9-5-2-6-10-12/h1-10H

InChI key

FOJFFKWENUWRFK-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jan Bergman et al.
The Journal of organic chemistry, 76(6), 1546-1553 (2011-02-24)
Tetraphosphorus decasulfide (P(4)S(10)) in pyridine has been used as a thionating agent for a long period of time. The moisture-sensitive reagent has now been isolated in crystalline form, and the detailed structure has been determined by X-ray crystallography. The thionating

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service