Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762393

Sigma-Aldrich

2-(4,4-Dimethyl-4,5-dihydro-2-oxazolyl)pyridine

97%

Synonym(s):

4,4-Dimethyl-2-(pyridin-2-yl)-4,5-dihydrooxazole, pymox-Me2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H12N2O
CAS Number:
Molecular Weight:
176.22
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

reaction suitability

reagent type: ligand

refractive index

n20/D 1.533

density

1.050 at 25 °C

storage temp.

2-8°C

SMILES string

CC1(C)COC(=N1)c2ccccn2

InChI

1S/C10H12N2O/c1-10(2)7-13-9(12-10)8-5-3-4-6-11-8/h3-6H,7H2,1-2H3

InChI key

ZANPCQHDEUORJP-UHFFFAOYSA-N

Application

Ligand for Ruthenium catalyzed C-H bond oxidation of arylalkanes.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chae S Yi et al.
Organic letters, 11(7), 1567-1569 (2009-02-28)
The cationic complex [(pymox-Me(2))RuCl(2)](+)BF(4)(-) was found to be a highly effective catalyst for the C-H bond oxidation reaction of arylalkanes in water. For example, the treatment of ethylbenzene (1.0 mmol) with t-BuOOH (3.0 mmol) and 1.0 mol % of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service