Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762083

Sigma-Aldrich

2,2-Difluoro-1,3-benzodioxole-5-carbonitrile

97%

Synonym(s):

2,2-Difluoro-5-cyano-1,3-benzodioxole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H3F2NO2
CAS Number:
Molecular Weight:
183.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

58-62 °C

functional group

fluoro
nitrile

SMILES string

FC1(F)Oc2ccc(cc2O1)C#N

InChI

1S/C8H3F2NO2/c9-8(10)12-6-2-1-5(4-11)3-7(6)13-8/h1-3H

InChI key

VMJNTFXCTXAXTC-UHFFFAOYSA-N

pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service