Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762075

Sigma-Aldrich

2-(Trifluoromethyl)cyclohexanol

mixture of cis and trans isomers, 97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11F3O
CAS Number:
Molecular Weight:
168.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

liquid

refractive index

n20/D 1.410

density

1.228 g/mL at 25 °C

functional group

fluoro
hydroxyl

SMILES string

OC1CCCCC1C(F)(F)F

InChI

1S/C7H11F3O/c8-7(9,10)5-3-1-2-4-6(5)11/h5-6,11H,1-4H2

Inchi Key

BGAGPUHWVOHHTP-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

140.0 °F

flash_point_c

60 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service