Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

762008

Sigma-Aldrich

Ethyl N-phenylformimidate

97%

Synonym(s):

N-(Ethoxymethylene)aniline, N-Phenylformimidic acid ethyl ester, N-Phenylmethanimidic acid ethyl ester, Ethyl isoformanilide

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$113.00

$113.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$113.00

About This Item

Empirical Formula (Hill Notation):
C9H11NO
CAS Number:
Molecular Weight:
149.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$113.00


Check Cart for Availability

Request a Bulk Order

Quality Level

assay

97%

form

liquid

refractive index

n20/D 1.528

density

1.006 g/mL at 25 °C

functional group

ether

SMILES string

CCO\C=N\c1ccccc1

InChI

1S/C9H11NO/c1-2-11-8-10-9-6-4-3-5-7-9/h3-8H,2H2,1H3/b10-8+

InChI key

DRDBNKYFCOLNQO-CSKARUKUSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

174.0 °F

flash_point_c

78.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service