761443
1,3,6,8(2H,7H)-Tetraone, 2,7-dicyclohexylbenzo[lmn][3,8]phenanthroline
98%
About This Item
Recommended Products
assay
98%
form
solid
mp
360-365 °C
λmax
382, 362, 243 nm in chloroform
semiconductor properties
N-type (mobility=6 cm2/V·s)
SMILES string
O=C1N(C2CCCCC2)C(=O)c3ccc4C(=O)N(C5CCCCC5)C(=O)c6ccc1c3c46
InChI
1S/C26H26N2O4/c29-23-17-11-13-19-22-20(26(32)28(25(19)31)16-9-5-2-6-10-16)14-12-18(21(17)22)24(30)27(23)15-7-3-1-4-8-15/h11-16H,1-10H2
InChI key
XWDVNWORIROXKG-UHFFFAOYSA-N
General description
Application
Features and Benefits
signalword
Warning
hcodes
pcodes
Hazard Classifications
Aquatic Acute 1
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Intrinsically stretchable active layers for organic field-effect transistors (OFET) are discussed. Polymer structural modification & post-polymerization modifications are 2 methods to achieve this.
Fabrication procedure of organic field effect transistor device using a soluble pentacene precursor.
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Thin, lightweight, and flexible electronic devices meet widespread demand for scalable, portable, and robust technology.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service