761397
(S)-(4,4′,6,6′-Tetramethoxybiphenyl -2,2′-diyl)bis(bis(3,5-dimethylphenyl )phosphine)
97%
Synonym(s):
(S)-Xyl-Garphos™
About This Item
assay
97%
form
solid
optical activity
[α]22/D -87.0, c = 0.5% in chloroform
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
>300 °C
functional group
phosphine
SMILES string
COc1cc(OC)c(c(c1)P(c2cc(C)cc(C)c2)c3cc(C)cc(C)c3)-c4c(OC)cc(OC)cc4P(c5cc(C)cc(C)c5)c6cc(C)cc(C)c6
InChI
1S/C48H52O4P2/c1-29-13-30(2)18-39(17-29)53(40-19-31(3)14-32(4)20-40)45-27-37(49-9)25-43(51-11)47(45)48-44(52-12)26-38(50-10)28-46(48)54(41-21-33(5)15-34(6)22-41)42-23-35(7)16-36(8)24-42/h13-28H,1-12H3
InChI key
IDNKFBDRZBLTFQ-UHFFFAOYSA-N
Legal Information
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service