Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

761117

Sigma-Aldrich

Dimethylformamidium trifluoromethanesulfonate

97%

Synonym(s):

Hydroxydimethylmethanaminium trifluoromethanesulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8F3NO4S
CAS Number:
Molecular Weight:
223.17
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

370-400 °C

functional group

amide
fluoro
triflate

SMILES string

C\[N+](C)=C/O.[O-]S(=O)(=O)C(F)(F)F

InChI

1S/C3H7NO.CHF3O3S/c1-4(2)3-5;2-1(3,4)8(5,6)7/h3H,1-2H3;(H,5,6,7)

InChI key

OTNRUIGCHNLSBT-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David E Jaramillo et al.
Nature materials, 19(5), 517-521 (2020-02-06)
Industrial processes prominently feature π-acidic gases, and an adsorbent capable of selectively interacting with these molecules could enable important chemical separations1-4. Biological systems use accessible, reducing metal centres to bind and activate weakly π-acidic species, such as N2, through backbonding

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service