Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760951

Sigma-Aldrich

Poly(ethylene glycol), 16 acetylene dendron, generation 3

average Mn 22,000

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

acetylene surface groups

form

solid

mol wt

PEG average Mn 20,000 (n~450; esterified with 16 acetylene groups)
PEG average Mn 20,000
average Mn 22,000

feature

generation 3

no. Surface Groups

16

transition temp

Tm 56-60 °C

functional group

alkyne

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biocompatible Dendritic Building Blocks for Advanced Biomedical Reasearch
Michael Malkoch and Andreas M Nystrom
Material Matters, 7(3), 47-49 null
Degradable Poly (ethylene glycol) hydrogels for 2 D and 3D cell culture
Andrea M Kasko
Material Matters, 6 (3), 67-75 null
Huiyuan Li et al.
Molecules (Basel, Switzerland), 18(8), 9797-9817 (2013-08-21)
Click chemistry is an efficient and chemoselective synthetic method for coupling molecular fragments under mild reaction conditions. Since the advent in 2001 of methods to improve stereochemical conservation, the click chemistry approach has been broadly used to construct diverse chemotypes

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service