Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760870

Sigma-Aldrich

Poly(ethylene glycol), 32 hydroxyl dendron, generation 4

average Mn 9,500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

description

hydroxyl surface groups

form

solid

mol wt

PEG average Mn 6,000 (n~130; esterified with 16 OH groups)
average Mn 9,500

feature

generation 4

no. Surface Groups

32

transition temp

Tm 42-48 °C

functional group

hydroxyl

Looking for similar products? Visit Product Comparison Guide

General description

Dendrimers with Poly(ethylene) glycol (PEG) core are widely used in drug delivery systems. These dendrimers are end functionalized with hydroxyl groups which increas its biocompatibility. PEG is used as the core since it is non-toxic, non-immunogenic with favorable pharmacokinetic and tissue distribution. They are also soluble in aqueous and many organic solvents.†††

Features and Benefits

Well-defined structure, high intrinsic drug loading capability, flexibility for tailored functionalization.[1]

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pharmaceutical and biomedical potential of PEGylated dendrimers.
Gajbhiye, V., Vijayaraj Kumar, P., Kumar Tekade, R., & Jain, N. K.
Current Pharmaceutical Design, 13(4), 415-429 (2007)
Chiara Clementi et al.
Molecular pharmaceutics, 8(4), 1063-1072 (2011-05-26)
Poly(ethylene glycol) (PEG) is the most popular polymer for protein conjugation, but its potential as carrier of low molecular weight drugs has been limited by the intrinsic low loading, owing to its chemical structure. In fact, only the two end
Marco D Giles et al.
Journal of the American Chemical Society, 130(44), 14430-14431 (2008-10-14)
At neutral pH, dendronized deep-cavity cavitands were shown to form supramolecular nanocapsules via assembly around a range of guest molecules.
Omayra L Padilla De Jesús et al.
Bioconjugate chemistry, 13(3), 453-461 (2002-05-16)
High molecular weight polymers (> 20 000 Da) have been widely used as soluble drug carriers to improve drug targeting and therapeutic efficacy. Dendritic polymers are exceptional candidates for the preparation of near monodisperse drug carriers due to their well-defined

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service