Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760846

Sigma-Aldrich

(R)-2,2′-Bis(bromomethyl)-1,1′-binaphthyl

97%

Synonym(s):

2,2′-Bis(bromomethyl)-1,1′-binaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H16Br2
CAS Number:
Molecular Weight:
440.17
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

optical activity

[α]22/D +160°, c = 1% in benzene

mp

181-185 °C

storage temp.

2-8°C

SMILES string

BrCc1ccc2ccccc2c1-c3c(CBr)ccc4ccccc34

InChI

1S/C22H16Br2/c23-13-17-11-9-15-5-1-3-7-19(15)21(17)22-18(14-24)12-10-16-6-2-4-8-20(16)22/h1-12H,13-14H2

InChI key

VUUUEDHFRJQSSY-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service