Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760420

Sigma-Aldrich

N,N,N′,N′-Tetrakis(3-methylphenyl)-3,3′-dimethylbenzidine

97% (HPLC)

Synonym(s):

3,3′-Dimethyl-N4,N4,N4′,N4′-tetra-m-tolyl-[1,1′-biphenyl]-4,4′-diamine, HMTPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H40N2
CAS Number:
Molecular Weight:
572.78
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97% (HPLC)

form

solid

mp

125-133 °C

SMILES string

Cc1cccc(c1)N(c2cccc(C)c2)c3ccc(cc3C)-c4ccc(N(c5cccc(C)c5)c6cccc(C)c6)c(C)c4

InChI

1S/C42H40N2/c1-29-11-7-15-37(23-29)43(38-16-8-12-30(2)24-38)41-21-19-35(27-33(41)5)36-20-22-42(34(6)28-36)44(39-17-9-13-31(3)25-39)40-18-10-14-32(4)26-40/h7-28H,1-6H3

InChI key

TZYPEOJJHBWOKE-UHFFFAOYSA-N

Application

This material is a novel high performance host material used for Organic Electroluminescent lighting devices for high efficiency and lifetimes. Internal efficiencies of 87% with external efficiencies of 20% were achieved.[1]

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C. Adachi et al.
Journal of Applied Physics, 90, 5048-5048 (2001)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service