Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

76032

Sigma-Aldrich

Palladium on calcium carbonate

5% Pd basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pd
MDL number:
UNSPSC Code:
12141733
eCl@ss:
38191001
PubChem Substance ID:

Quality Level

form

solid

autoignition temp.

>1120 °F

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

concentration

5% Pd

SMILES string

[Ca++].[Pd++].[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/2CH2O3.Ca.Pd/c2*2-1(3)4;;/h2*(H2,2,3,4);;/q;;2*+2/p-4

InChI key

XGLHAVVVUOEGIO-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Aqueous Heck coupling and hydroarylation
  • Reduction reactions
  • Hydrogenation reactions
Hydrogenation catalyst

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service