Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760285

Sigma-Aldrich

Indium(III) isopropoxide solution

5 % (w/v) in isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
In(OCH(CH3)2)3
CAS Number:
Molecular Weight:
292.08
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

≥99% trace metals basis

form

liquid

reaction suitability

core: indium

concentration

5 % (w/v) in isopropanol

refractive index

n20/D 1.383

bp

83-87 °C

density

0.808 g/mL at 25 °C

SMILES string

CC(C)O[In](OC(C)C)OC(C)C

InChI

1S/3C3H7O.In/c3*1-3(2)4;/h3*3H,1-2H3;/q3*-1;+3

InChI key

OVZUSPADPSOQQN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Sol-gel Precursor for the synthesis of indium oxide nanoparticles, nanocubes, thin films, indium tin oxide (ITO) films, showing interesting optical and conducting properties. Indium oxide and doped indium oxide nanoparticles and thin films finds applications in photovoltaics, gas sensors, example ozone, ethanol, hydrogen sensing etc. Also used for doping of transparent zinc oxide thin films.

pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Hoon Lee et al.
Journal of the American Chemical Society, 128(29), 9326-9327 (2006-07-20)
Hydrolysis of In(O-iPr)3 by 10 molar excess of water at 90 degrees C in a surfactant/solvent mixture of oleylamine/oleic acid/trioctylamine provides very small nanoparticles (<5 nm in diameter) of In(O)(OH). Subsequent in situ thermolysis of the formed In(O)(OH) nanoparticles at
Caruntu, D.; et al.
The Journal of Physical Chemistry C, 114, 4875-4875 (2010)

Articles

Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service