Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

760153

Sigma-Aldrich

Tin(IV) isopropoxide solution

10 % (w/v) in isopropanol/toluene, 99.99% trace metals basis

Synonym(s):

Isopropyl stannate(IV), Tetraisopropoxystannane, Tetraisopropoxytinisopropanol adduct, Tetraisopropyl stannate, Tin isopropoxide - isopropanol adduct, Tin tetraisopropoxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H28O4Sn
Molecular Weight:
355.06
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

Quality Level

assay

99.99% trace metals basis

form

liquid

reaction suitability

core: tin

concentration

10 % (w/v) in isopropanol/toluene

refractive index

n20/D 1.3942

bp

82.7 °C

density

0.8387 g/mL at 25 °C

SMILES string

CC(C)O[Sn](OC(C)C)(OC(C)C)OC(C)C

InChI

1S/4C3H7O.Sn/c4*1-3(2)4;/h4*3H,1-2H3;/q4*-1;+4

InChI key

CCTFOFUMSKSGRK-UHFFFAOYSA-N

Application

An important tin precursor for the synthesis of tin oxide nanoparticles; thin films; indium tin oxide (ITO) films; showing highly desired electrochemical and conducting properties. Doped tin oxide nanoparticles and thin films are widely studied as gas sensors.

signalword

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Central nervous system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

53.6 °F - closed cup

flash_point_c

12.0 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chen F.; Liu; M.
Chemical Communications (Cambridge, England), 18, 1829-1829 (1999)
Kumar; A.; et al.
Sens. Actuators, B155, 884-884 (2001)
Huang; J.; et al.
Chemistry of Materials, 17, 3513-3513 (2005)
Mengya Wang et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(45), 16195-16202 (2015-09-24)
A hierarchical fibrous SnO2 /carbon nanocomposite composed of fine SnO2 nanocrystallites immobilized as a thin layer on a carbon nanofiber surface was synthesized employing natural cellulose substance as both scaffold and carbon source. It was achieved by calcination/carbonization of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service