Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759988

Sigma-Aldrich

2-Chloro-8-fluoroquinoline

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5ClFN
CAS Number:
Molecular Weight:
181.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

assay

95%

form

solid

mp

75-85 °C

functional group

chloro
fluoro

SMILES string

Fc1cccc2ccc(Cl)nc12

InChI

1S/C9H5ClFN/c10-8-5-4-6-2-1-3-7(11)9(6)12-8/h1-5H

InChI key

CSPMWYOOKWBXRI-UHFFFAOYSA-N

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service