Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759945

Sigma-Aldrich

N-Formyl-Leu-OH

90%

Synonym(s):

N-Formyl-L-leucine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO3
CAS Number:
Molecular Weight:
159.18
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

assay

90%

form

solid

reaction suitability

reaction type: solution phase peptide synthesis

mp

138-145 °C

application(s)

peptide synthesis

storage temp.

−20°C

SMILES string

CC(C)C[C@H](NC=O)C(O)=O

InChI

1S/C7H13NO3/c1-5(2)3-6(7(10)11)8-4-9/h4-6H,3H2,1-2H3,(H,8,9)(H,10,11)/t6-/m0/s1

InChI key

HFBHOAHFRNLZGN-LURJTMIESA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

11 - Combustible Solids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service