Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759910

Sigma-Aldrich

1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole- 4,6(5H)-dione

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H17Br2NO2S
CAS Number:
Molecular Weight:
423.16
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
Product 759910 is not currently sold in your country. Contact Technical Service

Quality Level

assay

97%

form

solid

mp

109-114 °C

SMILES string

CCCCC(CC)CN1C(=O)c2c(Br)sc(Br)c2C1=O

InChI

1S/C14H17Br2NO2S/c1-3-5-6-8(4-2)7-17-13(18)9-10(14(17)19)12(16)20-11(9)15/h8H,3-7H2,1-2H3

InChI key

AOZLCBYWDXFKCJ-UHFFFAOYSA-N

General description

1,3-Dibromo-5-(2-ethylhexyl)-4H-thieno[3,4-c]pyrrole-4,6(5H)-dione is a thienopyrrolodione (TPD) based electron acceptor material (n-type semiconductor) which is used widely for organic photovoltaic (OPV) applications. They have very powerful electron withdrawing capability. TPD based conjugated polymers have exhibited a Power Conversion Efficiency (PCE) of as high as 7.3% in bulk heterojunction polymer solar cells.[1]

Application

  • This monomer is used in the synthesis of low band gap polymers for numerous OPV devices including bulk heterojunction solar cells[2].
  • Used as an electron acceptor material (n-type semiconductor) in Polymer Solar Cells.[3]

Features and Benefits

High electron withdrawing capability, deeper HUMO levels leading to high Voc. [1]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jong-jin Ha et al.
Chemistry, an Asian journal, 9(4), 1045-1053 (2014-01-31)
Two small molecules named BT-TPD and TBDT-TTPD with a thieno[3,4-c]pyrrole-4,6-dione (TPD) unit were designed and synthesized for solution-processed bulk-heterojunction solar cells. Their thermal, electrochemical, optical, charge-transport, and photovoltaic characteristics were investigated. These compounds exhibit strong absorption at 460-560 nm and low
Yuan; Mao-Chuan; Chou; et al.
Polymer, 52, 2792-2798 (2011)
Alternating Copolymers Consisting of Dialkoxylated Naphthalene along with Either Dithiophenylated 1, 4-Dioxopyrrolo-[3, 4c]-pyrrole or 5H-thieno [3, 4-c] pyrrole-4, 6-dione: Synthesis and Photo-Electrochemical Properties
Lim, Young Lae, et al.
Bull. Korean Chem. Soc., 34(1), 313-316 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service