Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759651

Sigma-Aldrich

1-Boc-6-oxa-1-azaspiro[3.3]heptane

95% (GC)

Synonym(s):

6-Oxa-1-azaspiro[3.3]heptane-1-carboxylic acid tert-butyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H17NO3
CAS Number:
Molecular Weight:
199.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

95% (GC)

form

solid

mp

33-40 °C

functional group

ether

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)N1CCC12COC2

InChI

1S/C10H17NO3/c1-9(2,3)14-8(12)11-5-4-10(11)6-13-7-10/h4-7H2,1-3H3

InChI key

FIFKIIBTYOOVJD-UHFFFAOYSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

No data available

flash_point_c

No data available


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service