Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759473

Sigma-Aldrich

m-Xylene-2,4,5,6-d4

98 atom % D, 99% (CP)

Synonym(s):

1,3-Dimethylbenzene-2,4,5,6-d4, Deuterium labeled m-xylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8D4H6
Molecular Weight:
110.19
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % D

assay

99% (CP)

bp

138-139 °C (lit.)

density

0.900 g/mL

mass shift

M+4

SMILES string

[2H]c1c([2H])c(C)c([2H])c(C)c1[2H]

InChI

1S/C8H10/c1-7-4-3-5-8(2)6-7/h3-6H,1-2H3/i3D,4D,5D,6D

InChI key

IVSZLXZYQVIEFR-LNFUJOGGSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

3 - Flammable liquids

wgk_germany

WGK 3

flash_point_f

77.0 °F

flash_point_c

25.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service