Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

759104

Sigma-Aldrich

3,3′-(4-Methyl-1,3-phenylene)bis(1,1-dibutylurea)-(tetrabutyl-d36)

≥98% D, ≥97% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25D36H8N4O2
Molecular Weight:
468.86
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98% D

assay

≥97% (CP)

form

solid

mol wt

~468.502 by atom % calculation

SMILES string

CC1=C(NC(N(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)C=C(NC(N(C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])[2H])=O)C

InChI

1S/C25H44N4O2/c1-6-10-16-28(17-11-7-2)24(30)26-22-15-14-21(5)23(20-22)27-25(31)29(18-12-8-3)19-13-9-4/h14-15,20H,6-13,16-19H2,1-5H3,(H,26,30)(H,27,31)/i1D3,2D3,3D3,4D3,6D2,7D2,8D2,9D2,10D2,11D2,12D2,13D2,16D2,17D2,18D2,19D2

InChI key

NMCJLWHQVDLBQO-UTRAKBEWSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service