758337
2,4-Bis[4-(N,N-diisobutylamino)-2,6-dihydroxyphenyl] squaraine
97%
About This Item
Recommended Products
assay
97%
form
solid
mp
295-305 °C
λmax
700 nm
orbital energy
HOMO 5.3 eV
LUMO 3.4 eV
OPV device performance
ITO/MoO3/SQ:C70 (1:5)/BCP/Al
- Short-circuit current density (Jsc): 13.7 mA/cm2
- Open-circuit voltage (Voc): 0.87 V
- Fill Factor (FF): 0.53
- Power Conversion Efficiency (PCE): 6.3 %
ITO/NPD/SQ/C60/BCP/Al
- Short-circuit current density (Jsc): 6.67 mA/cm2
- Open-circuit voltage (Voc): 0.82 V
- Fill Factor (FF): 0.55
- Power Conversion Efficiency (PCE): 3 %
ITO/SQ/C60/BCP/Al
- Short-circuit current density (Jsc): 7.13 mA/cm2
- Open-circuit voltage (Voc): 0.75 V
- Fill Factor (FF): 0.6
- Power Conversion Efficiency (PCE): 3.2 %
SMILES string
CC(C)CN(CC(C)C)c1cc(O)c([C]2[C+]([O-])[C]([C+]2[O-])c3c(O)cc(cc3O)N(CC(C)C)CC(C)C)c(O)c1
InChI
1S/C32H44N2O6/c1-17(2)13-33(14-18(3)4)21-9-23(35)27(24(36)10-21)29-31(39)30(32(29)40)28-25(37)11-22(12-26(28)38)34(15-19(5)6)16-20(7)8/h9-12,17-20,35-38H,13-16H2,1-8H3
InChI key
JRHMFCJQZUVUIN-UHFFFAOYSA-N
General description
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Solution-processed organic photovoltaic devices (OPVs) have emerged as a promising clean energy generating technology due to their ease of fabrication, potential to enable low-cost manufacturing via printing or coating techniques, and ability to be incorporated onto light weight, flexible substrates.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service