Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

758256

Sigma-Aldrich

2-Bromoisophthalaldehyde

97%

Synonym(s):

1-Bromo-2,6-diformylbenzene, 2-Bromobenzene-1,3-dialdehyde, 2-Bromobenzene-1,3-dicarboxaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5BrO2
CAS Number:
Molecular Weight:
213.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

139-144 °C

functional group

aldehyde
bromo

SMILES string

Brc1c(C=O)cccc1C=O

InChI

1S/C8H5BrO2/c9-8-6(4-10)2-1-3-7(8)5-11/h1-5H

InChI key

RZUSSKMZLHKMHU-UHFFFAOYSA-N

pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service