Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757993

Sigma-Aldrich

(S)-3-Amino-2-piperidone hydrochloride

95%

Synonym(s):

(S)-3-Amino-2-piperidinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11ClN2O
CAS Number:
Molecular Weight:
150.61
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

95%

form

solid

mp

180-203 °C (decomposition)

SMILES string

Cl.N[C@H]1CCCNC1=O

InChI

1S/C5H10N2O.ClH/c6-4-2-1-3-7-5(4)8;/h4H,1-3,6H2,(H,7,8);1H/t4-;/m0./s1

InChI key

NLAYLURYAOXTTE-WCCKRBBISA-N

Application

(S)-3-Amino-2-piperidone hydrochloride can be used as a building block for the synthesis of 5-hydroxy-2-[(3S)-2-oxo-3-piperidinyl]-1H-isoindole-1,3(2H)-dione by reacting with 4-hydroxyphthalic anhydride.[1] It is also used to prepare eukaryotic signaling peptide glorin and its synthetic analog glorinamide.[2]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Versatile synthesis of the signaling peptide glorin
Barnett R, et al.
Beilstein Journal of Organic Chemistry, 13(1), 247-250 (2017)
Synthesis, configurational stability and stereochemical biological evaluations of (S)-and (R)-5-hydroxythalidomides
Yamamoto T, et al.
Bioorganic & medicinal chemistry letters, 19(14), 3973-3976 (2009)
Ring-Expansion Reactions of Binaphthyl Azepines and Ferrocenophanes through Metal-Catalyzed [1, 2]-Stevens Rearrangements.
Harthong S, et al.
Synthesis, 45(15), 2070-2078 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service