Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

757756

Sigma-Aldrich

Poly(ethylene glycol) 2-aminoethyl ether biotin

average Mn 2,300

Synonym(s):

AminoPEG biotin, Biotin polyoxyethylene amine, Biotin-PEG, Biotin-PEG-amine, Biotinylated PEG, Polyoxyethylene ethylamine ether biotin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

PEG average Mn 2,000 (n~45)
average Mn 2,300

reaction suitability

reagent type: cross-linking reagent
reactivity: carboxyl reactive

mp

50-55 °C

Ω-end

amine

α-end

biotin

polymer architecture

shape: linear
functionality: heterobifunctional

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Applications:
  • bioconjugation
  • drug delivery

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Designing biomaterial scaffolds mimicking complex living tissue structures is crucial for tissue engineering and regenerative medicine advancements.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service