Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757578

Sigma-Aldrich

Potassium nitrate

anhydrous, 99.99% trace metals basis

Synonym(s):

Niter, Nitre, Nitric acid, potassium salt, Saltpeter

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
KNO3
CAS Number:
Molecular Weight:
101.10
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.99% trace metals basis

form

solid

impurities

≤200.0 ppm Trace Metal Analysis

mp

334 °C (lit.)

SMILES string

[K+].[O-][N+]([O-])=O

InChI

1S/K.NO3/c;2-1(3)4/q+1;-1

InChI key

FGIUAXJPYTZDNR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Very hygroscopic. Handle under inert conditions or dry box

pictograms

Flame over circle

signalword

Warning

hcodes

pcodes

Hazard Classifications

Ox. Sol. 3

Storage Class

5.1B - Oxidizing hazardous materials

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Valentin Loux et al.
BMC genomics, 16, 296-296 (2015-04-18)
Propionibacterium freudenreichii (PF) is an actinobacterium used in cheese technology and for its probiotic properties. PF is also extremely adaptable to several ecological niches and can grow on a variety of carbon and nitrogen sources. The aim of this work
Rose A Devasia et al.
The Journal of antimicrobial chemotherapy, 63(6), 1173-1178 (2009-03-31)
The aim of this study was to assess the sensitivity, specificity and time to results of mycobacterial growth indicator tube (MGIT) 960, microscopic observation drug susceptibility (MODS) assay and nitrate reductase assay (NRA) compared with the gold standard agar proportion
Jing Huang et al.
Applied and environmental microbiology, 80(1), 29-42 (2013-10-01)
Bacteria play important roles in mineral weathering and soil formation. However, few reports of mineral weathering bacteria inhabiting subsurfaces of soil profiles have been published, raising the question of whether the subsurface weathering bacteria are fundamentally distinct from those in
Changhong Yao et al.
Bioresource technology, 118, 438-444 (2012-06-22)
Microalgal starch is a potential feedstock for biofuel production. The effects of KNO(3) and MgSO(4) concentrations and light intensity on biomass and starch production by the marine microalga, Tetraselmis subcordiformis, were investigated. Under 200 μmol m(-2) s(-1) irradiance and sulfur-deprived
Wolfgang R Engelsberger et al.
The Plant journal : for cell and molecular biology, 69(6), 978-995 (2011-11-09)
Nitrogen is an essential macronutrient for plant growth and development. Inorganic nitrogen and its assimilation products control various metabolic, physiological and developmental processes. Although the transcriptional responses induced by nitrogen have been extensively studied in the past, our work here

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service