Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757500

Sigma-Aldrich

3-Chloro-L-alanine methyl ester hydrochloride

97%

Synonym(s):

β-Chloro-L-alanine methyl ester hydrochloride, (R)-Methyl 2-amino-3-chloropropanoate hydrochloride, (R)-Methyl 2-amino-3-chloropropionate hydrochloride, 3-Chloro- L-Ala-OMe hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H8ClNO2 · HCl
CAS Number:
Molecular Weight:
174.03
MDL number:
UNSPSC Code:
12352101
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

151.2-159.3 °C

functional group

amine
chloro
ester

storage temp.

2-8°C

SMILES string

Cl.COC(=O)[C@@H](N)CCl

InChI

1S/C4H8ClNO2.ClH/c1-8-4(7)3(6)2-5;/h3H,2,6H2,1H3;1H/t3-;/m0./s1

InChI key

POPBCSXDEXRDSX-DFWYDOINSA-N

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service