Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757403

Sigma-Aldrich

3-Amino-1-propanol-15N

98 atom % 15N, 98% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO(CH2)3NH2
Molecular Weight:
76.10
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
Pricing and availability is not currently available.

isotopic purity

98 atom % 15N

assay

98% (CP)

bp

184-187 °C

mp

10-12 °C

density

0.995 g/mL at 25 °C

mass shift

M+1

SMILES string

[15NH2]CCCO

InChI

1S/C3H9NO/c4-2-1-3-5/h5H,1-4H2/i4+1

InChI key

WUGQZFFCHPXWKQ-AZXPZELESA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

pictograms

CorrosionExclamation mark

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk_germany

WGK 1

flash_point_f

174.2 °F - closed cup

flash_point_c

79 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service