Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757381

Sigma-Aldrich

(S)-(−)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

(S)-(−)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole

97%

Synonym(s):

2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole, (6S)-4,5,6,7-Tetrahydro-2,6-benzothiazolediamine, (S)-4,5,6,7-Tetrahydro-2,6-benzothiazolediamine, (S)-4,5,6,7-Tetrahydrobenzo[d]thiazole-2,6-diamine, (S)-4,5,6,7-Tetrahydrobenzothiazole-2,6-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H11N3S
CAS Number:
Molecular Weight:
169.25
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

powder

mp

230.1-231.1 °C

storage temp.

2-8°C

SMILES string

N[C@H]1CCc2nc(N)sc2C1

InChI

1S/C7H11N3S/c8-4-1-2-5-6(3-4)11-7(9)10-5/h4H,1-3,8H2,(H2,9,10)/t4-/m0/s1

InChI key

DRRYZHHKWSHHFT-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(S)-(−)-2,6-Diamino-4,5,6,7-tetrahydrobenzothiazole can be used:
  • To prepare its sulfate monohydrate salt, which acts as a good material for nonlinear optical (NLO) applications.[1]
  • As a corrosion inhibitor for mild steel in acidic medium.[2]
  • As an intermediate in the synthesis of pramipexole, a dopamine D2 subfamily receptor agonist.[3]

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel scalable synthesis of pramipexole
Z?ivec M, et al.
Organic Process Research & Development, 14(5), 1125-1129 (2010)
Structural characterization, vibrational study, NLO and DFT calculations of a novel organic sulfate monohydrate templated with (S)-(-)-2, 6-diammonium-4, 5, 6, 7-tetrahydrobenzothiazole
Barhoumi A, et al.
Journal of Molecular Structure, 1128, 544-551 (2017)
Exploration of (S)-4, 5, 6, 7-tetrahydrobenzo [d] thiazole-2, 6-diamine as feasible corrosion inhibitor for mild steel in acidic media
Manivel A, et al.
Journal of Environmental Chemical Engineering, 2(1), 463-470 (2014)

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service