Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757292

Sigma-Aldrich

9,10-Dichlorooctafluoroanthracene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14Cl2F8
CAS Number:
Molecular Weight:
391.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

solid

mp

166-173 °C

SMILES string

ClC1=C2C(C(F)=C(F)C(F)=C2F)=C(Cl)C3=C1C(F)=C(F)C(F)=C3F

InChI

1S/C14Cl2F8/c15-5-1-2(8(18)12(22)11(21)7(1)17)6(16)4-3(5)9(19)13(23)14(24)10(4)20

InChI key

QQLRAWFMOWCSBU-UHFFFAOYSA-N

Application

Used in the synthesis of n-type Organic Semiconductors.[1]

pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John F Tannaci et al.
The Journal of organic chemistry, 72(15), 5567-5573 (2007-06-26)
9,10-Dichlorooctafluoroanthracene (1) was synthesized from commercially available tetrafluorophthalic acid by an optimized solution-phase route. To establish 1 as a synthon for n-type organic semiconductors, the compound was reacted with phenylboronic acid under modified Suzuki-Miyaura coupling conditions to generate octafluoro-9,10-diphenylanthracene (7)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service