Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757217

Sigma-Aldrich

1-Phenylisoquinoline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H11N
CAS Number:
Molecular Weight:
205.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

97%

form

powder

mp

90-95 °C

SMILES string

c1ccc(cc1)-c2nccc3ccccc23

InChI

1S/C15H11N/c1-2-7-13(8-3-1)15-14-9-5-4-6-12(14)10-11-16-15/h1-11H

InChI key

LPCWDYWZIWDTCV-UHFFFAOYSA-N

General description

This material can be used as a ligand to synthesize a variety of OLED dopants.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen-Yao Zhang et al.
European journal of medicinal chemistry, 178, 401-416 (2019-06-16)
This work mainly introduces the synthesis and characterization of three iridium(III) complexes [Ir(ppy)2(adppz)](PF6) (Ir-1), [Ir(bzq)2(addpz)](PF6) (Ir-2) and [Ir(piq)2(adppz)](PF6) (Ir-3). The complexes are more cytotoxic than cisplatin against tumor cell lines such as SGC-7901, A549, HeLa, Eca-109, HepG2 and BEL-7402. The
Menghan Zang et al.
Talanta, 202, 259-266 (2019-06-07)
A group of luminescent dinuclear Ir(III) complexes were synthesized to study the application possibility as luminescent G-quadruplex (G4) sensors. According to the results, the dinuclear Ir(III) complexes 1-5 were assessed as selective G4 sensors. Among them, complex 5 not only

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service