Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757195

Sigma-Aldrich

2-(4-Ethyl-2-pyridyl)-1H-imidazole

98%

Synonym(s):

2-(4-Ethyl-2-pyridinyl)-1H-imidazole, 4-Ethyl-2-(1H-imidazol-2-yl)pyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N3
Molecular Weight:
173.21
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

98%

form

powder or crystals

mp

88-92 °C

SMILES string

CCC1=CC=NC(C2=NC=CN2)=C1

InChI

1S/C10H11N3/c1-2-8-3-4-11-9(7-8)10-12-5-6-13-10/h3-7H,2H2,1H3,(H,12,13)

InChI key

OUJMVVWGIGNJER-UHFFFAOYSA-N

Application

This material can be used as a ligand to synthesize a variety of OLED dopants.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service