Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

757160

Sigma-Aldrich

4,4′-Bis(3-ethyl-N-carbazolyl)-1,1′-biphenyl

99% (HPLC)

Synonym(s):

4,4′-Bis(3-ethyl-9H-carbazol-9-yl)-1,1′-biphenyl, 9,9′-(1,1′-Biphenyl)-4,4′-diylbis(3-ethylcarbazole), Ethyl-CBP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H32N2
Molecular Weight:
540.70
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

assay

99% (HPLC)

form

solid

mp

243-248 °C

SMILES string

CCc1ccc2n(-c3ccc(cc3)-c4ccc(cc4)-n5c6ccccc6c7cc(CC)ccc57)c8ccccc8c2c1

InChI

1S/C40H32N2/c1-3-27-13-23-39-35(25-27)33-9-5-7-11-37(33)41(39)31-19-15-29(16-20-31)30-17-21-32(22-18-30)42-38-12-8-6-10-34(38)36-26-28(4-2)14-24-40(36)42/h5-26H,3-4H2,1-2H3

InChI key

QITOABKVTZWLGX-UHFFFAOYSA-N

Application

This modified CBP material can be used as a high performance host material analagous to CBP. The ethyl group modifies the HOMO LUMO Energy levels for the tuning of OLED devices.

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service