Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756873

Sigma-Aldrich

2-Bromothiazole-4-carboxaldehyde

Synonym(s):

2-Bromo-4-formylthiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H2BrNOS
CAS Number:
Molecular Weight:
192.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

solid

mp

126-132 °C

functional group

aldehyde
bromo

storage temp.

2-8°C

SMILES string

Brc1nc(C=O)cs1

InChI

1S/C4H2BrNOS/c5-4-6-3(1-7)2-8-4/h1-2H

InChI key

MNQVIZWWCRPZOK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service