Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756717

Sigma-Aldrich

Bisphenol F, mixture of acetates and propionates

Synonym(s):

4,4′-Methylenebisphenol, bis(mixed acetates and propionates), EC-298

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002

form

liquid

refractive index

n20/D 1.549

density

1.143 g/mL at 25 °C

Storage temp.

2-8°C

General description

Bisphenol F (BPF) is an epoxy based resin that can be used as an engineering polymer. It has a low viscosity, high thermal stability and chemical resistance. It can be used in anticorrosive coatings and epoxy based adhesives. It can be cured by using amine and anhydride based hardeners. It has an epoxy value of 0.549 mol/100g.[1][2][3]

Application

BPF is a resin that can be used in the development of high strength materials in aircrafts and electronic products. It can form a variety of engineering plastics for industrial applications.[4][5]
Epoxy curative with good thermal properties.
  • Stable
  • Low viscosity
  • Does not impede free radical cure
  • Thermoset adhesives
  • Curative for epoxy/(meth) acrylate hybrids
  • Hybrid epoxy/free radical thermosets

Pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Curing kinetics and thermal property characterization of the bisphenol-F epoxy resin and phthalic anhydride system.
Gao J, et al.
Polymer International, 51(12), 1422-1427 (2002)
Flame-retardant electrical conductive nanopolymers based on bisphenol F epoxy resin reinforced with nano polyanilines.
Zhang X, et al.
ACS Applied Materials & Interfaces, 5(3), 898-910 (2013)
Effective functionalization of carbon nanotubes for bisphenol F epoxy matrix composites.
Wang Z, et al.
Materials Research, 15(4), 510-516 (2012)
Novel surface modified molecularly imprinted polymer focused on the removal of interference in environmental water samples for chromatographic determination.
Watabe Y, et al.
Journal of Chromatography A, 1073(1-2), 363-370 (2005)
Cure kinetics, gelation, and glass transition of a bisphenol F epoxide.
O'Brien DJ and White SR
Polym. Eng. Sci., 43(4), 863-874 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service