Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

756571

Sigma-Aldrich

Sodium yttrium oxyfluoride, ytterbium and erbium doped

greener alternative

red up-conversion phosphor, 99% trace metals basis

Synonym(s):

Red up-conversion phosphor, Ytterbium and erbium doped sodium yttrium oxyfluoride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
NaY1.54Yb0.40Er0.06F5O
Molecular Weight:
350.15
UNSPSC Code:
12352300
NACRES:
NA.23

assay

99% trace metals basis

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

particle size distribution

1-5 Ī¼m (d50)

fluorescence

Ī»ex 940-980 nm (Excitation source)

greener alternative category

General description

Red up-conversion phosphor
Type BPP-1071

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

>1292.0 °F

flash_point_c

> 700 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hua Zhang et al.
Chemical communications (Cambridge, England), 47(3), 979-981 (2010-11-17)
We report a five-fold overall enhancement of upconversion emission in NaYF(4) : Yb/Er nanocrystals when coupled with gold island films. Spectroscopic studies show that the enhancement factors are highly dependent on the exact spectral positions and excitation power density, with a largest
Mai, H.-X.; et al.
The Journal of Physical Chemistry C, 11, 13721-13721 (2007)
Yang, X.; et al.
J. Mater. Sci., 42, 7042-7042 (2007)
Cui, L.; et al.
Rare Metals, 26, 276-276 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service