756555
Sodium yttrium fluoride, ytterbium and erbium doped
green up-conversion phosphor, 99% trace metals basis
Synonym(s):
Erbium sodium ytterbium yttrium fluoride, Green up-conversion phosphor, Ytterbium and erbium doped sodium yttrium fluoride
About This Item
Recommended Products
assay
99% trace metals basis
form
powder
greener alternative product characteristics
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
particle size
1-5 μm (d50)
fluorescence
λem 940-980 nm (UV Excitation Source)
greener alternative category
General description
Application
Storage Class
13 - Non Combustible Solids
wgk_germany
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service