Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

756539

Sigma-Aldrich

Strontium aluminate, europium and dysprosium doped

greener alternative

long persistent green phosphor

Synonym(s):

Aluminum dysprosium europium strontium oxide, Europium and dysprosium doped strontium aluminum oxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Sr0.95Eu0.02Dy0.03Al2O4
CAS Number:
Molecular Weight:
209.11
UNSPSC Code:
12352300
NACRES:
NA.23

assay

98% trace metals basis

form

powder

composition

Sr0.95Eu0.02Dy0.03Al2O4

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

particle size

-230 mesh

mp

>1200 °C

fluorescence

λem Green

greener alternative category

General description

Strontium aluminate, europium and dysprosium doped is a luminescent compound, which can be used in persistent luminous paints, inks, and ceramics. It can be synthesized by a variety of methods such as sol-gel, hydrothermal synthesis, solid-state reactions, and combustion reactions. It can also be utilized in the preparation of efficient bifunctional structural layers for the fabrication of quantum dot dye-sensitized solar cells (DSSCs).[1][2][3]

Application

Long persistent green phosphor
SrAl2O4:Eu2+,Dy3+ can be as a green phosphor for applications in cathode ray tubes,[4] smart textiles,[5] and dye sensitized solar cells (DSSCs).[6]

Other Notes

Persist Time (min): 9500 ± 10%

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and mechanoluminescent property of (Eu2+, Dy3+)-co-doped strontium aluminate phosphor by soft mechanochemistry-assisted solid-state method
Lv H, et al.
Journal of Luminescence, 209, 129-140 (2019)
Influence of calcining temperature on photoluminescence and triboluminescence of europium-doped strontium aluminate particles prepared by sol- gel process
Liu Y and Xu C
The Journal of Physical Chemistry B, 107(17), 3991-3995 (2003)
Enhanced performance of cadmium selenide quantum dot-sensitized solar cells by incorporating long afterglow europium, dysprosium co-doped strontium aluminate phosphors
Sun H, et al.
Journal of Colloid and Interface Science, 416(6), 81-85 (2014)
Smart textile framework: Photochromic and fluorescent cellulosic fabric printed by strontium aluminate pigment
Khattab TA, et al.
Carbohydrate Polymers, 195(17), 143-152 (2018)
Enhanced performance of cadmium selenide quantum dot-sensitized solar cells by incorporating long afterglow europium, dysprosium co-doped strontium aluminate phosphors
Sun H, et al.
Journal of Colloid and Interface Science, 416(17), 81-85 (2014)

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service