Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

756407

Sigma-Aldrich

Barium nitride

powder, -60 mesh, 99.7% trace metals basis, Sr: <0.8%

Synonym(s):

Tribarium dinitride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba3N2
CAS Number:
Molecular Weight:
439.99
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.23

assay

99.7% trace metals basis

form

powder

particle size

-60 mesh

SMILES string

[Ba]=N[Ba]N=[Ba]

InChI

1S/3Ba.2N

InChI key

ZNGBRDVORXINKX-UHFFFAOYSA-N

Application

Barium nitride is a ceramic that finds use as a dopant in optical materials such as white emitting LEDs. Ba3N2 is also used as a synthetic precursor for ternary metal nitrides and as a flux growth medium for GaN.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

supp_hazards

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bao, H.Q.; et al.
Journal of Crystal Growth, 310, 2955-2955 (2008)
Berger, U.; Schnick, W.
J. Alloy Compounds, 206, 179-179 (1994)
Bonghyun Lee et al.
ACS combinatorial science, 13(2), 154-158 (2011-02-01)
We employed a solid-state combinatorial chemistry technique to screen 4 ternary phosphor systems: (Sr,Ca,Ba)(2)Si(5)N(8):Eu(2+), (Sr,Ca,Mg)(2)Si(5)N(8):Eu(2+), (Sr,Mg,Ba)(2)Si(5)N(8):Eu(2+), and (Ca,Ba,Mg)(2)Si(5)N(8):Eu(2+). The current pure nitride-based system did not allow for the use of conventional liquid solution-based high-throughput experimentation, so that a specially designed

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service