Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756199

Sigma-Aldrich

N,N-Diisobutylaniline

95%

Synonym(s):

N,N-Bis(2-methylpropyl)benzenamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H23N
CAS Number:
Molecular Weight:
205.34
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

assay

95%

form

liquid

refractive index

n20/D 1.516

density

0.899 g/mL at 25 °C

SMILES string

CC(C)CN(CC(C)C)c1ccccc1

InChI

1S/C14H23N/c1-12(2)10-15(11-13(3)4)14-8-6-5-7-9-14/h5-9,12-13H,10-11H2,1-4H3

InChI key

LMOOEMRNWCISOX-UHFFFAOYSA-N

Application

Readily forms adduct with borane, which is further applied in hydroboration and reduction of several carbonyl bearing functionalities and nitriles.

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

10 - Combustible liquids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Brown, H. C.; Kanth, J. V. B.; Zaidlewicz, M.
The Journal of Organic Chemistry, 63, 5154-5163 (1998)
Brown, H. C.; et al.
The Journal of Organic Chemistry, 64, 6263-6274 (1999)
Brown, H. C.; Zaidlewicz, M.; Dalvi, P. V.
Organometallics, 17, 4202-4202 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service