Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

756091

Sigma-Aldrich

4,6-Dichloro-2-(methylthio)pyrimidine-5-carbonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3Cl2N3S
CAS Number:
Molecular Weight:
220.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

assay

97%

form

solid

mp

106-110 °C

SMILES string

CSc1nc(Cl)c(C#N)c(Cl)n1

InChI

1S/C6H3Cl2N3S/c1-12-6-10-4(7)3(2-9)5(8)11-6/h1H3

InChI key

GLOOTGZVAHKTJS-UHFFFAOYSA-N

pictograms

Skull and crossbonesCorrosion

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral - Skin Corr. 1B

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Letters, 50, 370-370 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service