Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755974

Sigma-Aldrich

Holmium

dendritic pieces, purified by distillation, 99.99% trace rare earth metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ho
CAS Number:
Molecular Weight:
164.93
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

99.99% trace rare earth metals basis

form

dendritic pieces

purified by

distillation

density

8.8 g/mL at 25 °C (lit.)

SMILES string

[Ho]

InChI

1S/Ho

InChI key

KJZYNXUDTRRSPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Schellhorn et al.
Optics letters, 38(4), 504-506 (2013-03-05)
We report the first observation to our knowledge of room-temperature continuous-wave laser operation on the (5)I(7)→(5)I(8) transition of Ho(3+) ions in a KY(3)F(10) single crystal. Using a Tm-doped silica fiber laser operating at 1938 nm as a pump source, a
Y Nedjadi et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 70(9), 1990-1996 (2012-03-17)
The work presented here is a new and precise measurement of the half-life of (166m)Ho by determining the activity concentration, using an ionisation chamber calibrated for this nuclide, and measuring the number of (166m)Ho atoms using multi-collector inductively coupled plasma
Nicolas Wartenberg et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(10), 3477-3482 (2013-01-29)
The bis-tetrazolate-pyridine ligand H(2)pytz sensitises efficiently the visible and/or near-IR luminescence emission of ten lanthanide cations (Pr, Nd, Sm, Eu, Tb, Dy, Ho, Er, Tm, Yb). The Ln(III) complexes present sizeable quantum yields in both domains with a single excitation
Tomonori Hu et al.
Optics letters, 37(11), 2145-2147 (2012-06-05)
We report an efficient Q-switched Ho(3+)Pr(3+)-doped fluoride fiber laser, producing a peak power of 77 W, with pulse width of 78 ns. A slope efficiency of 20% with respect to the launched pump power was achieved. A TeO(2) acousto-optic modulator
Alexander Hemming et al.
Optics express, 21(4), 4560-4566 (2013-03-14)
We report the highest power operation of a resonantly cladding-pumped, holmium-doped silica fibre laser. The cladding pumped all-glass fibre utilises a fluorine doped glass layer to provide low loss cladding guidance of the 1.95 µm pump radiation. The operation of

Articles

A significant limiting factor for wearable electronics and wireless sensors is the finite amount of energy that can be stored in on-board batteries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service