Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755788

Sigma-Aldrich

2-[8-(3-Hexyl-2,6-dioctylcyclohexyl)octyl]pyromellitic diimide oligomer, maleimide terminated, lower viscosity

Synonym(s):

BMI-5000P, (1,2-Bis(octylmaleimide)-3-octyl-4-hexyl)cyclohexyl oligomer, BMI-9000P, Imide-extended bismaleimide oligomer

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

transition temp

Tg 85-90 °C

Looking for similar products? Visit Product Comparison Guide

General description

·Low cross-link density
·Non-tacky
·Film-forming
·Maleimide functional oligomer
·Cures to a tough thermoset
·Additive to enhance toughness in thermoset compositions
Well-suited for film adhesives and thermally resistant adhesives
Viscosity(50% in toluene): 1,000 cps @ 25 C

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service