Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755761

Sigma-Aldrich

Diaqua[1,3-bis(diphenylphosphino)propane]palladium(II) bis(tetrafluoroborate)

95%

Synonym(s):

[Pd(dppp)(H2O)2](BF4)2

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H30O2P2Pd · 2BF4
CAS Number:
Molecular Weight:
728.50
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

assay

95%

form

solid

reaction suitability

core: palladium
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst

mp

175-200 °C (decomposition)

storage temp.

2-8°C

SMILES string

O.O.[Pd++].F[B-](F)(F)F.F[B-](F)(F)F.C(CP(c1ccccc1)c2ccccc2)CP(c3ccccc3)c4ccccc4

InChI

1S/C27H26P2.2BF4.2H2O.Pd/c1-5-14-24(15-6-1)28(25-16-7-2-8-17-25)22-13-23-29(26-18-9-3-10-19-26)27-20-11-4-12-21-27;2*2-1(3,4)5;;;/h1-12,14-21H,13,22-23H2;;;2*1H2;/q;2*-1;;;+2

InChI key

RJHRHADURPIBJJ-UHFFFAOYSA-N

Application

Versatile Palladium precatalyst for cyclization reactions.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cationic Palladium Complex-Catalyzed Diastereoselective Tandem Annulation of 2-Iminoarylboronic Acids with Substituted Alkynes: Enantioselective Synthesis of Aminoindene Derivatives by Double Asymmetric Induction
Yu, X.; Lu, X.
Advanced Synthesis & Catalysis, 353, 2805-2813 (2011)
Xufen Yu et al.
Organic letters, 11(19), 4366-4369 (2009-09-02)
Cationic palladium complex [(dppp)Pd(H(2)O)(2)](2+)(BF(4)(-))(2) catalyzed tandem annulation reactions of 2-formylarylboronic acids and allenoates produce the indenol derivatives diastereo- and enantioselectively in a highly efficient way.
Xiuling Han et al.
Organic letters, 12(1), 108-111 (2009-12-09)
High chemoselectivity for the synthesis of two kinds of substituted coumarins controlled by the counteranions of the cationic palladium catalysts is described. The asymmetric version of the reaction for the synthesis of 3-alkylidene dihydrocoumarins is realized with high enantioselectivity.
Xufen Yu et al.
The Journal of organic chemistry, 76(15), 6350-6355 (2011-07-13)
The 1-benzoxepine derivatives were synthesized conveniently by cationic palladium-catalyzed [5 + 2] annulation reaction of 2-acylmethoxyarylboronic acids with allenoates in high yields. This annulation involves the intramolecular nucleophilic addition to ketones without the formation of π-allylpalladium species.
Kun Shen et al.
Organic letters, 14(7), 1756-1759 (2012-03-28)
Cationic Pd(II)-catalyzed enantioselective arylative cyclization of alkyne-tethered enals or enones initiated by carbopalladation of alkynes was developed without the necessity of a redox system.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service