Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

755699

Sigma-Aldrich

Erbium(III) chloride

anhydrous, beads, -10 mesh, 99.99% trace metals basis

Synonym(s):

Erbium trichloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ErCl3
CAS Number:
Molecular Weight:
273.62
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

grade

anhydrous

assay

99.99% trace metals basis

form

beads

particle size

-10 mesh

mp

774 °C (lit.)

density

4.1 g/mL at 25 °C (lit.)

SMILES string

Cl[Er](Cl)Cl

InChI

1S/3ClH.Er/h3*1H;/q;;;+3/p-3

InChI key

HDGGAKOVUDZYES-UHFFFAOYSA-K

Looking for similar products? Visit Product Comparison Guide

pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L Sudheendra et al.
Chemistry of materials : a publication of the American Chemical Society, 23(11), 2987-2993 (2011-06-29)
A metal shell was used in this study to provide significant enhancement of the up-converted emission from cubic NaYF(4) nanoparticles, creating a valuable composite material for labeling in biology and other applications - use of the cubic form of the
Yi, G.; et al.
Chemistry of Materials, 23, 2729-2729 (2011)
A P Russell et al.
Analytical biochemistry, 164(1), 181-189 (1987-07-01)
A method of measuring the volumetric magnetic susceptibility, in which magnetically labeled cells or other particles are suspended in a paramagnetic solution of known susceptibility over the poles of a magnet, is presented. If the cells are more magnetic than
Yue Wang et al.
Frontiers in chemistry, 8, 238-238 (2020-05-07)
We report a new sensor for the specific detection of lead ions (Pb2+) in contaminated water based on fluorescence resonance energy transfer (FRET) between upconversion nanoparticles (UCNPs) as donors and gold nanoparticles (Au NPs) as receptors. The UCNPs modified with
M Zborowski et al.
Journal of general microbiology, 138(1), 63-68 (1992-01-01)
A trivalent lanthanide ion, erbium (Er3+), has been used in combination with a magnetic separation technique to isolate seven bacterial species from suspensions in 0.9% saline. Erbium has an exceptionally high atomic magnetic moment of 9.3 Bohr magnetons, and following

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service